Tubing Rack, 10-Channel for 1/4" OD Tubing, Non-Adhesive Back, 10/Pk
Representative image only.
Mfr # PCTR250-102 – Item # EW-06432-03
An easy solution to secure and route multiple tubing lines
PRODUCT OPTIONS
$16.75USD / Pkg of 10
SPECS & DESCRIPTION
Specifications & Description
  • Number of Channels10
  • For Tubing OD (in)1/4
  • For Tubing OD (mm)6.35
  • DescriptionTubing Rack, 10-Channel for 1/4" OD Tubing, Non-Adhesive Back, 10/Pk
More About this Item
These tube racks offer a simple and economical method for organizing your tubing lines. The 3/16" and 1/4" sizes can be easily separated down to single holders by hand for increased flexibility and custom tubing arrangements. The 3/16" and 1/4" sizes are available with adhesive fastener, and all sizes are available with through holes for secure mounting (mounting screws are not included). Standard racks are black polyethylene; 3/16" and 1/4" sizes are available in other colors. Contact Cole-Parmer for more information.
REVIEWS
REVIEW SNAPSHOT®
by PowerReviews
NoneTubing Rack, 10-Channel for 1/4" OD Tubing, Non-Adhesive Back, 10/Pk
4.0
(based on 1 review)
Reviewed by 1 customer
Displaying review 1
Back to top
4.0
Just what I was looking for
By 
from TX
About Me Academic/Professional Researcher
Verified Buyer
Pros
  • Easy To Use
Cons
  • Innacurate
Best Uses
  • In The Field
  • In The Lab
Comments about None Tubing Rack, 10-Channel for 1/4" OD Tubing, Non-Adhesive Back, 10/Pk:
The channels are under sized as indicated. The 1/4" measures only 0.2" so tubing with a true 1/4" OD is severly pinched. I would order the next size up.
Displaying review 1
Back to top