Tubing Rack, 5-Channel for 1/2" OD Tubing, Non-Adhesive Back, 10/Pk
Representative image only.
Item # EW-06432-05
An easy solution to secure and route multiple tubing lines
PRODUCT OPTIONS
$51.00USD / Pkg of 10
SPECS & DESCRIPTION
Specifications & Description
  • Number of Channels5
  • For Tubing OD (in)1/2
  • For Tubing OD (mm)12.7
  • DescriptionTubing Rack, 5-Channel for 1/2" OD Tubing, Non-Adhesive Back, 10/Pk
More About this Item
These tube racks offer a simple and economical method for organizing your tubing lines. The 3/16" and 1/4" sizes can be easily separated down to single holders by hand for increased flexibility and custom tubing arrangements. The 3/16" and 1/4" sizes are available with adhesive fastener, and all sizes are available with through holes for secure mounting (mounting screws are not included). Standard racks are black polyethylene; 3/16" and 1/4" sizes are available in other colors. Contact Cole-Parmer for more information.