Flux F460EX-UL 120V Explosion-proof electric motor, 9000 rpm, 120 VAC, 60 Hz

Representative image only.
Flux Mfr # F460EX-UL 120V – Item # EW-07076-10

$1,478.00USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Max Motor RPM (rpm)9000
  • Power (VAC)120
  • Power (Hz)60
  • CE Regulatory ApprovalYes
  • UL(US) Regulatory ApprovalYes
  • DescriptionExplosion-proof electric motor, 9000 rpm, 120 VAC, 60 Hz

More About this Item

Explosion-proof electric motors are rated for Class 1, Groups C and D hazardous environments and for pumping flammable liquids. Manual overload reset feature eliminates the danger of automatic restarting.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat