John Guest Speedfit Ball Valve, straight, 1/4" OD speedfit x 1/4" OD speedfit
Representative image only.
John Guest Mfr # PPSV040808W – Item # EW-07391-04
Quickly and easily connect and disconnect valve
  • Constructed of durable polypropylene (PP)
PRODUCT OPTIONS
$12.10USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Connection Type1/4" OD Speedfit to 1/4" OD Speedfit
  • Max Pressure (PSI)150
  • Max Pressure (bar)10.3
  • Max Temperature (° F)150
  • Max Temperature (° C)65
  • Wetted Materialsfood-grade polypropylene, EPDM, and stainless steel
  • DescriptionSpeedfit Ball Valve, straight, 1/4" OD speedfit x 1/4" OD speedfit
Key Features
  • Constructed of durable polypropylene (PP)
More About this Item
Food-grade polypropylene housing is specifically made for use with potable water. Quickly disconnect and reconnect your water lines with these Speedfit valves.