This item is now discontinued. Please browse similar items
Measurement Specialties 406 400 Series Thermistor Liquid-Immersion Probe; 1/8" Dia
Representative image only.
Measurement Specialties Mfr # 406 – Item # EW-08436-00
SPECS & DESCRIPTION
Specifications & Description
  • Probe TypeThermistor
  • Min Temperature (° F)-40
  • Max Temperature (° F)302
  • Min Temperature (° C)-40
  • Max Temperature (° C)150
  • Time Constant2.5 seconds
  • Connector TypePhono plug
  • Sheath Length (in)4.5
  • Sheath Diameter (in)0.125
  • Description400 Series Thermistor Liquid-Immersion Probe; 1/8" Dia
More About this Item
Probes are accurate to +/- 0.1 deg C from 0 to 70 deg C. Electrically isolated probes come with a nondetachable 10-ft lead with phone plug (except as noted).
Water-resistant junctions between probe and lead should not be immersed; detachable leads are not water resistant. Note: Time constant is estimate time required to reach 63.2% of a new reading. To determine how long it will take to reach 99% of a new reading multiply the time constant by five.
1/4" or 6.350 mm phono plug connection