Brinkmann 023210900 Vertical Top-loading Autoclave 23 L, 220 VAC

Representative image only.
Brinkmann Mfr # 023210900 – Item # EW-10780-17
Automatic temperature and pressure regulation cuts heating time up to 50%
  • Easy-to-read, bright blue LCD panel highlights pressure, temperature, time, and process alerts
  • Low-water sensor prevents activation of sterilization cycle when there is insufficient water in the chamber or reservoir
  • Four preloaded cycles simplify sterilization of liquids and wrapped or unwrapped instruments
  • 1 year warranty

SPECS & DESCRIPTION

Specifications & Description

  • Chamber Diameter (in)9.88
  • Chamber Depth (in)13.81
  • Max Temperature (° F)280
  • Max Temperature (° C)138
  • Max Pressure (PSI)30
  • Width (in)19 11/16
  • Height (in)26 13/16
  • Chamber construction316 Stainless Steel
  • DisplayLCD
  • Power (VAC)208
  • Power (Hz)60
  • Power (amps)10
  • DescriptionVertical Top-loading Autoclave 23 L, 220 VAC
  • Warranty1 year
1 Year WarrantyUL Listed.

Key Features

  • Easy-to-read, bright blue LCD panel highlights pressure, temperature, time, and process alerts
  • Low-water sensor prevents activation of sterilization cycle when there is insufficient water in the chamber or reservoir
  • Four preloaded cycles simplify sterilization of liquids and wrapped or unwrapped instruments
  • 1 year warranty

More About this Item

Simple one-touch sterilization and drying make these vertical sterilizers ideal for sterilizing glass and plastic containers, media, buffers, solid instruments, and hazardous waste materials. The dual temperature and pressure control cuts heating time up to 50% by systematically increasing the chamber pressure and temperature. Throughout the heating phase, the chamber and sample temperature reach equilibrium in half the time of other autoclaves. Rapid cooling model decreases chamber cool-down time by up to 90%, offering quicker access to your sample and the ability to perform more sterilization cycles daily.
Sterilizers offer added user safety and security with a heat-resistant door, audible and visual cycle interruption alerts, and a double safety-locking device to prevent the door from

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat