This item is now discontinued. We recommend the following as suggested replacements: EW-12148-24
Thermo Scientific Haake 440-0051 Programmable 250°C Bath, 230 VAC, 50/60 Hz
Representative image only.
Thermo Scientific Haake Mfr # 440-0051 – Item # EW-12202-23
Powerful pumping capacity — up to 24 liters per minute
  • Turbulence reduction system adjusts pump flow for open bath applications
  • Store up to three or four fixed-temperature settings
  • 2 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionProgrammable 250°C Bath, 230 VAC, 50/60 Hz
  • Warranty2 year
Two year warranty
Key Features
  • Turbulence reduction system adjusts pump flow for open bath applications
  • Store up to three or four fixed-temperature settings
  • 2 year warranty
More About this Item
Use these heated circulating baths for controlling temperatures in smaller external systems, such as density meters, viscometers, photometers or refractometers. Bath opening allows samples to be inserted into the circulators' own bath for internal heating.
Programmable Circulating Baths are ideal for a full range of internal and external control applications. Powerful
pressure and suction pumps provide constant pressure and flow rate over the full temperature range. Intelligent FuzzyStar® control technology automatically adjusts pump speed to liquid viscosity for accurate, stable, and efficient temperature management. Stores one program and four user-defined fixed
temperature settings. Assign menu options to function keys. RS-232 connection permits remote operation. Large display provides operating steps, temperature readings, and fault descriptions in six selectable languages—German, English, French, Italian, Spanish, and Japanese. Durable stainless steel tanks are easy to clean.