Thermo Scientific Haake 426-3001 Immersion Circulators, Digital control, RS-232, 230 VAC, 50/60 Hz
Representative image only.
Thermo Scientific Haake Mfr # 426-3001 – Item # EW-12206-47
Temperature control for baths up to 50 liters
  • Turbulence reduction system reduces pump speed to 10 L/min for open baths
  • 2 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionImmersion Circulators, Digital control, RS-232, 230 VAC, 50/60 Hz
  • Warranty2 year
Two year warrantyISO9001 Certified Supplier
Key Features
  • Turbulence reduction system reduces pump speed to 10 L/min for open baths
  • 2 year warranty
More About this Item
Swivel-mounted nozzle ensures thorough mixing and even temperature distribution. Attach circulator to any bath wall with a thickness of up to 1'' (25-mm) using screw clamp (included). Digital models display the reason whenever the circulator automatically cuts off. Real temperature adjustment ensures accurate display. RS-232 output on models 12206-42 and -47 lets you connect the circulator to a PC. Order optional tap water cooling coil 12200-51 separately.