This item is now discontinued. Please browse similar items

Bioscan HI-425-145 Microplate reader, liquid scintillation and luminescence

Representative image only.
Bioscan Mfr # HI-425-145 – Item # EW-13094-08
Up to five technologies in one unit!
  • Read absorbance, luminescence, liquid scintillation, fluorescence intensity (FI), and time-resolved fluorescence (TRF)
  • Designed to read 24-, 96- and 384-well plates

SPECS & DESCRIPTION

Specifications & Description

  • Light Sourcexenon flash lamp-closed loop
  • Height (in)11 3/4
  • Width (in)14 1/2
  • Length (in)15 13/64
  • Power (VAC)110 to 240
  • Power (Hz)50/60
  • DescriptionMicroplate reader, liquid scintillation and luminescence

Key Features

  • Read absorbance, luminescence, liquid scintillation, fluorescence intensity (FI), and time-resolved fluorescence (TRF)
  • Designed to read 24-, 96- and 384-well plates

More About this Item

These microplate readers are compact in size and save valuable bench space. An internal microprocessor with easy-to-use software accesses all instrument functions. Choose between single or combined function units. All models can have additional functions added at a later time
For advanced data analysis order optional DeltaSoft PC software. DeltaSoft software includes nine curve fitting programs, statistical analysis of sample concentration calculations, detailed kinetic analysis and report templates that are Excel™ compatible.
The Liquid Scintillation Counter has preset modes for the most commonly used radioisotopes (3H, 14C, 35S, 33P and 125I). Use for scintillation proximity assays (SPA), flash plate assays, cell proliferation studies, radioimmunoassays (RIA) and immunoradiometric assays (IRMA).
The Fluorescence Reader is ideal for both short life-time fluorophores and long life-time fluorescence using Fluorescence Intensity (FI), and Time-Resolved Fluorescence (TRF) modes. Applications include DNA hybridization studies, binding studies, enzyme assays, ligand receptor binding assays, cytotoxicity and immunoassays.
The Luminometer measures both “glow” and “flash” type reagents. Use for ATP detection, reporter gene assays, luminescence immunoassays, and toxicology. Luminescence function requires an injector (sold separately).
The Absorbance Reader has a wavelength range from 240 to 1000 nm. Typical applications include DNA quantification, colorimetric assays and enzyme assays.

Specifications:
Detectors: photon counting photomultiplier and photo diode
Optical type: direct illumination (fiberless optics)
Light source: xenon flash lamp-closed loop
Fluorescence sensitivity:
Fluorescence intensity (FI): <2 fmol
Fluorescence polarization (FP): 5mP
Time-resolved fluorometry (TRF): <100amol Eu
Excitation: 230-620 nm
Emission: 360-630 nm (700nm optional)
Dynamic range: >5 decades
Luminescence sensitivity: <1 amol ATP/well, >6 decades dynamic range
Liquid Scintillation Counting Efficiency: 30% for 3-H, 50% for I-125, and 80% for C-14 in a 96-well plate
Absorbance::
Linearity: 2%
Dynamic range: 3.0 OD
Wavelength range: 220 to 900nm

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat