This item is now discontinued. Please browse similar items
0-8247-0175-5 Food Packaging: Principles and Practice
Representative image only.
Mfr # 0-8247-0175-5 – Item # EW-15819-01
SPECS & DESCRIPTION
Specifications & Description
  • ISBN0-8247-0175-5
  • AuthorG.L. Robertson
  • Published Date1998
  • Number of Pages688
  • DescriptionFood Packaging: Principles and Practice
More About this Item
A comprehensive guide to food packaging technologies, this text examines the aspects of packaging that are relevant to the processing, preservation, distribution, and marketing of a particular food and the areas of food science and technology that influence the packaging process. Covers everything from types of deteriorative reactions to packaging requirements of different food groups.