Accumet XL 30 benchtop conductivity meter only

Representative image only.
Accumet Mfr # CPXL30A – Item # EW-19551-20
Measure conductivity, resistivity, salinity, TDS, and temperature
  • Uses 2 and 4-cell conductivity probes
  • Customizable features — adapt for specific applications
  • 3 year warranty

SPECS & DESCRIPTION

Specifications & Description

  • Meter TypeBenchtop
  • Display typeLED
  • Conductivity Accuracy±0.5% FS + 1 digit
  • Cell ConstantK = 0.1, 1.0, or 10
  • TDS Resolution0.1 ppm; 0.1 ppt
  • TDS Accuracy±0.5% of FS + 1 digit
  • Min Salinity0 to 90 ppt
  • Salinity Resolution0.001 or 0.01 ppt
  • Salinity Accuracy±0.5% of reading
  • Min Resistivity3 Ω to 100 MΩ (4 ranges)
  • Resistivity Accuracy±0.5% ±1 digit
  • Temperature Accuracy±0.2°C
  • Temperature compensationAutomatic or manual
  • Temperature Coefficient0 to 10%/°C
  • InterfaceEthernet, RS-232, and USB
  • Data Logging (points)250 data sets
  • Power (Hz)50 or 60
  • Width (cm)23.495
  • Height (cm)16.51
  • Width (in)9 1/4
  • Length (in)3 1/2
  • Height (in)6 1/2
  • Descriptionbenchtop conductivity meter only
  • Warranty3 year
3 Year WarrantyISO9001:2000 Certified Supplier

Key Features

  • Uses 2 and 4-cell conductivity probes
  • Customizable features — adapt for specific applications
  • 3 year warranty

More About this Item

Advanced XL 30 Meter measures conductivity, resistivity, salinity, TDS and temperature. Meter features a 4-1⁄2"L x 6"H (11.4 x 15.2 cm), scratch-resistant, color LCD touchscreen with 3⁄4"H (1.9 cm) characters. Displays current measured value, stability indicator, standards in use with time/date stamp, temperature, and more.

Windows CE.net based operating system with on-screen operational prompts, recognizable icons, and context specific help screens make this meter easy to set up and use. Selectable temperature coefficients, display units, resolution, screen configurations, buffer sets, and print configurations provide flexibility for a variety of applications.

Meter accepts 2- or 4-cell conductivity probes with constants of K = 0.1, 1.0, and 10. Order the convenient meter kit that includes an ATC cell (K = 1.0), or meter only with your choice of probes listed below. Choose from probes with built-in ATC or order ATC probe 94000-62 and non-temperature compensated cells.

XL 30 Meter includes: electrode support arm and bracket, and SMPS 100/240 VAC, 50/60 Hz power supply.

XL 30 Meter kit includes: two-cell conductivity probe (K = 1.0) with built-in ATC probe (19550-64), electrode support arm and bracket, and SMPS 100/240 VAC, 50/60 Hz power supply.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat