This item is now discontinued. Please browse similar items

Horiba 352734-1 D-54 pH/Conductivity Meter

Representative image only.
Horiba Mfr # 352734-1 – Item # EW-19600-20
Large LCD shows temperature and pH, mV, or conductivity simultaneously
  • Self diagnostic function ensures reliable measurements
  • Nonvolatile memory stores all calibration data and up to 300 measurements

SPECS & DESCRIPTION

Specifications & Description

  • Meter TypePortable
  • Display typeLCD
  • Min pH (pH)0
  • Max pH (pH)14
  • pH Resolution0.01/0.1
  • pH Accuracy±0.01 pH
  • Buffer RecognitionDIN, NIST, and USA
  • Min mV (mV)-1999
  • Max mV (mV)1999
  • mV Resolution1 mV
  • mV Accuracy±1mV 1 digit
  • Conductivity Accuracy±0.5% FS 1 LSD
  • Cell ConstantAdjustable
  • Temperature Accuracy±0.1°C ±1 digit
  • Temperature compensationAutomatic
  • InterfaceRS-232
  • WaterproofYes
  • Width (cm)7.62
  • Height (cm)17.78
  • Width (in)3
  • Length (in)2 1/4
  • Height (in)7
  • DescriptionD-54 pH/Conductivity Meter

Includes

soft-side case with strap, and batteries. Order electrodes separately.

Key Features

  • Self diagnostic function ensures reliable measurements
  • Nonvolatile memory stores all calibration data and up to 300 measurements

More About this Item

Waterproof meters feature IP67-rated housing for field or lab use. Download data using RS-232 capability — order cable 99591-70 separately. Order digital conductivity simulator 19600-30 to calibrate meter without having to use liquid standards. D-54 meter measures pH, temperature, ORP, and conductivity.

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat