This item is now discontinued. View the direct replacement: EW-23022-71 or browse similar items

Buchi 23122A020 Rotavapor R-215 Professional Rotary Evaporator, Diagonal; 115V

Representative image only.
Buchi Mfr # 23122A020 – Item # EW-23122-02
Renowned systems with advanced features for ease of use
  • Digital display for both temperature and rotation speed
  • Multiple glassware configurations to meet your needs

SPECS & DESCRIPTION

Specifications & Description

  • Display TypeDigital
  • Condenser TypeDiagonal
  • Min Speed (rpm)20
  • Max Speed (rpm)280
  • Speed Resolution1 rpm
  • Glass TypeUncoated
  • Joint Size24/40
  • Lift MechanismQuick-action jack
  • Bath TypeWater
  • Bath Capacity (L)4
  • Min Temperature (° C)20
  • Max Temperature (° C)180
  • Vacuum SealPTFE
  • Power (VAC)115
  • Power (watts)1300
  • Power (Hz)60
  • Length (in)21 21/32
  • Width (in)22 41/64
  • Height (in)16 11/32
  • Length (cm)55
  • Width (cm)58
  • Height (cm)42
  • DescriptionRotavapor R-215 Professional Rotary Evaporator, Diagonal; 115V

Includes

quick-action jack, 1-liter glass flasks (one receiving and one evaporating), condenser, a 5-ft power cord with three-prong plug, water bath with digital display, and professional vacuum controller.

Key Features

  • Digital display for both temperature and rotation speed
  • Multiple glassware configurations to meet your needs

More About this Item

Use these evaporators with flasks from 50 mL to 4 liters. Chemical-resistant PTFE and long-life seals provide practically maintenance-free operation and use. Sparkless, brushless motor provides safe, quiet operation with constant torque from 20 to 280 rpm.

The water/oil bath has a temperature range from 20 to 180°C. Bath has been designed with energy efficiency and safety in mind. Insulating housing prevents contact with hot surfaces; high alloy stainless steel pan is easy to clean. Heating coils are located beneath the surface of the pan to minimize maintenance and simplify cleaning. Use only uncoated flasks when using as an oil bath. Bath features wireless contacts for both power supply and communications, adding to maximum safety for both the unit and user. In the event of a power failure or fault in the bath, the flask is automatically raised to prevent overheating of the product. Each unit features digital display for bath temperature.

Choose from a wide variety of glassware to meet your needs. The diagonal condenser is used for basic distillations and is ideal for where height is limited. The vertical condenser is also used for basic distillations, however takes up less bench space. The vertical assembly also features a connection for a vapor temperature sensor. The cold-trap condenser is ideal for maximizing the condensation due to the low temperatures and no additional cooling water necessary. For distillations needing reflux, choose from either vertical or cold-trap condenser with shutoff. All glassware (except for evaporating flasks) is available with Plastic + Glas coating to minimize breakage and safety concerns.

Note: Direct exposure of Plastic + Glas glassware to chlorinated solvents and aggressive reagents should be avoided. After extensive use, coating on glassware may wear and loosen from surface, increasing risk of injury.

Use these evaporators with flasks from 500 mL to 4 liters. Chemical-resistant PTFE and long-life seals provide practically maintenance-free operation and use. Sparkless, brushless motor provides safe, quiet operation with constant torque from 20 to 280 rpm.
The diagonal condenser is used for basic distillations, and is ideal for where height is limited.
The professional vacuum controller allows you to automate your distillation and program your rotary evaporator for easy repetition of your parameters. Vacuum controller allows you to regulate the vacuum to a specified set-point, control quick-action jack and rotation of the rotary evaporator, and provides USB interface for data transfer. In addition, the timer function interrupts the process after a set time.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat