Masterflex Fitting, Polycarbonate, Four-Way Stopcock, Male Luer Lock, Non-Sterile; 10/PK
Representative image only.
Masterflex Item # EW-30600-04
Regulate flow without any leaking
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
PRODUCT OPTIONS
$40.75USD / Pkg of 10
BUY MORE & SAVE
  • 1-9$40.75USD / Pkg of 10
  • 10-49$36.68USD / Pkg of 10
  • 50-99$34.64USD / Pkg of 10
  • 100+$32.60USD / Pkg of 10
SPECS & DESCRIPTION
Specifications & Description
  • Outlet PortMale Luer Lock
  • Inlet PortFemale Luer
  • Wetted MaterialsPolycarbonate
  • Flow PatternFour-Way
  • DescriptionStopcocks with Luer Connections; 4-way; male lock, Non-sterile
Key Features
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Always in stock—most materials and configurations available to ship same day
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Four-way stopcock/flow diverter with male luer lock connections
  • HDPE handle rotates 360°
More About this Item
Choose stopcocks with either a male luer lock or a male luer slip. Handle rotates 90° on one-way stopcocks, 180° on three-way stopcocks, and 360° on four-way stopcocks. All have PC body and HDPE stem.