Overlook Industries Disposable filler nozzle, SS needle and polycarbonate base, 1/8
Representative image only.
Overlook Industries Mfr # OVCP-1/8-1-D – Item # EW-30619-07
Reduce filler line downtime and risk of cross-contamination
  • Lower costs for cleaning, sterilization, and validation
  • Provide enhanced flexibility in batch scheduling and product development
  • Dimensional tolerances of +/-0.05% for assured repeatability
PRODUCT OPTIONS
Free Certificate of Comliance
$216.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Fitting MaterialPolycarbonate
  • Sterilization MethodAutoclave, ethylene oxide (ETO), or gamma
  • Connection TypeBarb
  • Barb Size / Tubing Id (Port 1) (in)1/8
  • Fitting Length (in)4
  • DescriptionDisposable filler nozzle, SS needle and polycarbonate base, 1/8
Certificate of Compliance
Key Features
  • Lower costs for cleaning, sterilization, and validation
  • Provide enhanced flexibility in batch scheduling and product development
  • Dimensional tolerances of +/-0.05% for assured repeatability
More About this Item
These nozzles combine the ease of single-use plastic components with the accuracy and reliability of stainless steel filler needles. Tight dimensional tolerances help to ensure precision in your dispensing applications. Nozzles feature a 316L stainless steel needle and a base with hose barb adapter. Materials meet USDA and USP Class VI requirements, and is FDA compliant. All filler nozzles are sterilizable by ethylene oxide, autoclaving, or gamma irradiation (note: polycarbonate will turn a greenish-yellow hue when gamma irradiated).