Masterflex Fitting, HDPE, Straight, Hose Barb to Threaded Adapter, 3/16" ID x 1/2" NPT(M); 10/PK
Representative image only.
Masterflex Item # EW-31702-18
Quick and easy assembly
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
PRODUCT OPTIONS
$8.70USD / Pkg of 10
BUY MORE & SAVE
  • 1-9$8.70USD / Pkg of 10
  • 10-49$7.83USD / Pkg of 10
  • 50-99$7.40USD / Pkg of 10
  • 100+$6.96USD / Pkg of 10
SPECS & DESCRIPTION
Specifications & Description
  • Fitting MaterialHDPE
  • Sterilization MethodEthylene oxide (ETO) only
  • Number Of Ports2
  • Connection TypeBarb x NPT(M)
  • Thread size NPT(M) (port 2) (in)1/2
  • Barb Size / Tubing Id (Port 1) (in)3/16
  • Flow PatternStraight
  • DescriptionThreaded adapter Male 1/2 x 3/16" pack of 10
Key Features
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Always in stock—most materials and configurations available to ship same day
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Connection Type: 3/16" ID x 1/2" NPT(M)
More About this Item
Available in a variety of materials and configurations to meet your need. Adapts from a NPT male thread to a hose barb.