Masterflex Variable-Area Flowmeter, Direct Reading, Graph-Style, Aluminum Housing, 10 scfm Air
Representative image only.
Masterflex Item # EW-32206-04
Easily read graph-style measurements directly
  • Direct-reading flowmeters feature a calibrated scale for a specific medium - no need to convert readings to get flow rate│Use with air for laboratory and industrial applications
PRODUCT OPTIONS
$295.60USD / Each
$272.00USD / Each
for an additional
$272.00USD / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • Accuracy±2% full-scale
  • Repeatability±1%
  • Material HousingAluminum
  • Max Operating Temperature (° F)240
  • Max Operating Temperature (° C)115.5
  • DescriptionGraph-style aluminum flowmeter 1/4"NPT, 1000 psi, 1-10 scfm
Key Features
  • Direct-reading flowmeters feature a calibrated scale for a specific medium - no need to convert readings to get flow rate│Use with air for laboratory and industrial applications
More About this Item
Use for high pressure industrial applications—install in any configuration, even next to a bend in your pipeline. A calibrated retention spring eliminates the need to mount flowmeters vertically or run straight lengths of pipe prior to or after the meter. These rugged flowmeters are relatively insensitive to shock and vibrations—the spring, piston assembly, and internal magnet compensate for rough process environments.