Cole-Parmer Compact low-flow switch for liquids; SPST normally open, 0.05 GPM
Representative image only.
Cole-Parmer Item # EW-32778-31
$160.70USD / Each
$296.70USD / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • Max Operating Temperature (° F)212
  • Max Operating Temperature (° C)100
  • DescriptionCompact low-flow switch for liquids; SPST normally open, 0.05 GPM
More About this Item
These normally open switches are specifically designed for reliable operation in clean air and water applications. Select from gas or liquid flow switches. Use a 50-µm filter to protect your switch. Switches include 18" (45.7 cm) L leads.