Pyrex 2094-1L 2094 Large Volume Class B Glass Burette, 1000 mL; 1/Cs

Representative image only.
Pyrex Mfr # 2094-1L – Item # EW-34542-34
Handle more fluid
  • Maintain fluid integrity within burette (buret) with dust cap
  • Designed with a 1000 mL capacity

PRODUCT OPTIONS

$248.00USD / Case of 1

$76.00USD / Each

Time to recalibrate? Click on the calibration service to view details, then order.
Often bought with…

SPECS & DESCRIPTION

Specifications & Description

  • Capacity (mL)1000
  • Graduation Interval (mL)10
  • Tolerance5
  • Description2094 Large Volume Class B Glass Burette, 1000 mL; 1/Cs

Includes

dust cover.

Key Features

  • Maintain fluid integrity within burette (buret) with dust cap
  • Designed with a 1000 mL capacity

More About this Item

Burettes (burets) are ideal for those requiring a larger capacity — up 1000 mL. These Class B dispensing burettes are equiped with a PTFE 2-mm bore stopcock plug to reduce freezing and lubricant contamination. Barrel has a microfinish for precision fit and the tips have the accuracy to provide required drainage rate. Each burette has a durable colored scale and dust cover.

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat