This item is now discontinued. View the direct replacement: EW-35413-00 or browse similar items

Oakton benchtop pH/conductivity/TDS meter with probes, 110/220 VAC

Representative image only.
Oakton Item # EW-35610-10
Easily toggle between pH, conductivity, and TDS measurements at the touch of a button!
  • Seperate probes to suit a variety of applications
  • Conductivity and TDS autoranging chooses the best resolution for each measurement

SPECS & DESCRIPTION

Specifications & Description

  • Meter TypeBenchtop
  • Display typeLCD
  • Min pH (pH)-2
  • Max pH (pH)16
  • pH Resolution0.01/0.1
  • pH Accuracy±0.01 pH
  • Buffer RecognitionDIN, NIST, and USA
  • CalibrationUp to 5 points
  • Conductivity Resolution0.01 or 0.1 μS; 0.001, 0.01, or 0.1 mS
  • Conductivity Accuracy±1% FS
  • Min TDS0 to 200 ppt
  • TDS Accuracy±1% of reading
  • Temperature compensationAutomatic or manual
  • Power (VAC)115
  • Power (Hz)60
  • Width (cm)22.86
  • Height (cm)6.0325
  • Width (in)9
  • Height (in)2
  • Descriptionbenchtop pH/conductivity/TDS meter with probes, 110/220 VAC

Key Features

  • Seperate probes to suit a variety of applications
  • Conductivity and TDS autoranging chooses the best resolution for each measurement

More About this Item

The pH/conductivity/TDS 510 meter features pH auto-buffer recognition, selectable °F/°C, HOLD, and water-resistant membrane keypad. Previous calibration points, electrode slope and offset, and conductivity/TDS cell constants are stored in memory for quick and easy retrieval.

Meter includes: electrode holder arm and universal power supply

Meter with probes adds: 59001-70 double-junction pH electrode with BNC connector & 35608-50 conductivity/TDS/temperature probe.

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat