Oakton Flexible Benchtop Electrode Holder
Representative image only.
Oakton Item # EW-35617-50
Keeps electrodes in position
  • Weighted base pivots 360° for added convenience
  • 1 year warranty
$122.50USD / Each
Include Electrodes Today!
Oakton® pH Epoxy-Body Electrodes with ATC
ITEM(S) ON SALE
$132.30 - $162.90USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionFlexible Benchtop Electrode Holder
  • Warranty1 year
1 Year Warranty
Key Features
  • Weighted base pivots 360° for added convenience
  • 1 year warranty
More About this Item
Holds up to four standard size electrodes (14-mm dia max) and one ATC probe. Weighted 6'' (15.2 cm) diameter base lets you pivot holder 360° for added convenience. Flexible arm lets you move electrode side to side or up and down while keeping electrode at a constant vertical angle.