This item is now discontinued. View the direct replacement: EW-35419-03.

Oakton pH 510 meter with "All-in-One" pH/Temp electrode, 110/220 VAC

Representative image only.
Oakton Item # EW-35619-03
Advanced features at an economical price!
  • Store and recall 100 readings in memory
  • Measures pH, temperature, and relative mV
  • Recorder output allows you to create hard copies

SPECS & DESCRIPTION

Specifications & Description

  • Display typeLCD
  • Min pH (pH)0
  • Max pH (pH)14
  • pH Resolution0.01/0.1
  • pH Accuracy±0.01 pH
  • Min mV (mV)-1999
  • Max mV (mV)1999
  • mV Resolution0.1 mV for ±199.9 mV; 1 mV beyond ±199.9 mV
  • mV Accuracy±0.2 mV or 0.05%, whichever is greater
  • Temperature compensationAutomatic or manual
  • Power (VAC)110 / 220
  • Power (Hz)50 or 60
  • Width (cm)17.78
  • Height (cm)6.0325
  • Width (in)7
  • Height (in)2 3/8
  • DescriptionpH 510 meter with "All-in-One" pH/Temp electrode, 110/220 VAC

Includes

110/220 VAC power supply

Key Features

  • Store and recall 100 readings in memory
  • Measures pH, temperature, and relative mV
  • Recorder output allows you to create hard copies

More About this Item

These easy-to-use, microprocessor-based meters feature hold, ATC, recorder output, and electrode diagnostics. The convenient pull-out quick reference card keeps procedures handy at all times. The large, dual-display LCD provides excellent visibility—even from a distance. Meters accept electrodes with a BNC connector. pH 510 meters measures pH, temperature, and relative mV. Automatic temperature compensation maintains reading accuracy even in fluctuating temperatures. Five-point pH calibration with automatic buffer recognition for both NIST (pH 1.68,4.01, 6.86, 9.18, 12.45) and USA (pH 1.68, 4.01, 7.00, 10.01,12.45) pH buffers, makes for easy calibration no matter which requirement you need to meet. Create hard copies of your data by using the recorder output.

ACCESSORIES

REVIEWS

REVIEW SNAPSHOT®

by PowerReviews
Oakton<reg> pH 510 meter with "All-in-One" pH/Temp electrode, 110/220 VAC
 
4.0

(based on 1 review)

Reviewed by 1 customer

Displaying review 1

Back to top

 
4.0

I would also add an electrode holder

By 

from Hyde Park Mass.

About Me Academic/Professional Researcher

Verified Buyer

Pros

  • Accurate
  • Durable
  • Easy To Use
  • It has a lot of features
  • The cost is very low
  • The unit is user friendly
  • Well Calibrated
  • Works Properly

Cons

  • No electrode holder

Best Uses

  • Back-up to my portables
  • In The Lab

Comments about Oakton<reg> pH 510 meter with "All-in-One" pH/Temp electrode, 110/220 VAC:

I like the display ,The over all ease of use and the multiple functions packed into one unit at a really affordable price.

Displaying review 1

Back to top

Q&A

Do you have questions about this product?

Get answers from real customers and in-house experts with AnswerBox.

1 Question | 1 Answer
Displaying question 1
  • Q:

    What is the part number for the electrode that comes with this unit? I need a replacement. Thanks!
    Asked on 12/17/2010 by Emily from Cedar Rapids, IA

    1 answer

    • STAFF EXPERT

      A:

      The electrode that comes with this product is part number: 35811-71.

      Answered on 12/21/2010 by Jack from Technical Support
Displaying question 1

Do you have a question about this product? 

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat