Digi-Sense Acorn Temperature 4 Thermistor Thermometer with Boot, with Out Probe
Representative image only.
Digi-Sense Item # EW-35626-00
Splashproof sealed membrane keypad makes cleanup easy
  • Minimum and Maximum display
  • Push-button temperature offset calibration adjustment for fine-tuning factory settings
  • HOLD button freezes measured reading
  • Switchable between °C and °F
  • Nonvolatile memory retains data — even when the battery is removed.
  • Auto power-off 17 minutes after last key press
  • Protective rubber boot features a built-in stand.
  • 3 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Meter Measurement TypeThermistor
  • Number Of Probe Channels1
  • Types Of Probes AcceptedThermistor
  • Min Temperature (° F)-40
  • Max Temperature (° F)257
  • Min Temperature (° C)-40
  • Max Temperature (° C)125
  • Accuracy±0.4°F (±0.2°C)
  • Resolution0.1°
  • Low Battery IndicatorYes
  • Display Type0.875\" Display, 3.5-digit LCD
  • Height (in)1 1/2
  • Depth (in)2 3/4
  • Battery Life200 hours
  • BatteryAAA Battery (4)
  • DescriptionAcorn Temperature 4 Thermistor Thermometer with Boot, with Out Probe
  • Warranty3 year
3 Year WarrantyConformite Europeenne
Key Features
  • Minimum and Maximum display
  • Push-button temperature offset calibration adjustment for fine-tuning factory settings
  • HOLD button freezes measured reading
  • Switchable between °C and °F
  • Nonvolatile memory retains data — even when the battery is removed.
  • Auto power-off 17 minutes after last key press
  • Protective rubber boot features a built-in stand.
  • 3 year warranty
More About this Item
Ideal for routine temperature measurements, these thermometers offer the convenience and functionality of expensive meters at an economical price. Max/min key momentarily displays maximum or minimum value; or use with HOLD key to update display only on new maximum (minimum) value. Additional features include under range and over range error messages and manual user calibration.