This item is now discontinued. View the direct replacement: EW-35634-55 or browse similar items

Oakton SaltTestr 11 pocket salinity tester

Representative image only.
Oakton Item # EW-35662-52
Ideal for spot checking NaCl-based samples.
  • Replaceable sensor extends instrument life
  • IP67-rated waterproof housing
  • Factory calibrated against NaCl standards

SPECS & DESCRIPTION

Specifications & Description

  • Meter TypeTester
  • Display typeLCD
  • Calibration1 point
  • Min Salinity0 to 10 ppt
  • Salinity Resolution0.1 ppt; 0.01%
  • Temperature Accuracy±0.9°F (±0.5°C)
  • Temperature compensationAutomatic
  • WaterproofYes
  • Battery4 x A76 batteries (included)
  • Battery life (hr)150
  • Width (cm)3.81
  • Height (cm)3.81
  • Descriptionpocket salinity tester

Includes

four 1.5V button cell batteries.

Key Features

  • Replaceable sensor extends instrument life
  • IP67-rated waterproof housing
  • Factory calibrated against NaCl standards

More About this Item

Push-button calibration provides easy and precise field calibration. Hold function freezes measurement for viewing and recording.

ACCESSORIES

REVIEWS

REVIEW SNAPSHOT®

by PowerReviews
Oakton SaltTestr 11 pocket salinity tester
 
4.0

(based on 1 review)

Reviewed by 1 customer

Displaying review 1

Back to top

(1 of 1 customers found this review helpful)

 
4.0

SaltTestr fantastic tool

By 

from McKinney, TX

About Me Academic/Professional Researcher

Verified Reviewer

Pros

  • Accurate
  • Easy To Use
  • Well Calibrated
  • Works Properly

Cons

  • Breaks Easily

Best Uses

  • In The Field

Comments about Oakton SaltTestr 11 pocket salinity tester:

I use it to test salt levels in swimming pools, it's fast, easy to use and accurate. The temperature reading lets you know when the reading has stabilized. I use it to calibrate and troubleshoot the installed equipment as well. It could come with a more durable case. Make sure you lubricate the body O-rings with silicone lube when you change the batteries, I lost my first one to leakage that way.

Displaying review 1

Back to top

Q&A

Do you have questions about this product?

Get answers from real customers and in-house experts with AnswerBox.

1 Question | 1 Answer
Displaying question 1
  • Q:

    Can this be used to measure salt content of soil/dirt?

    Thanks,
    Asked on 1/31/2017 by Dave from CA

    1 answer

    • STAFF EXPERT

      A:

      This can be used to measure the salt content of soil/dirt but the soil/dirt will need to be dissolved in water. This will not be able to take a measurement of a dried sample.

      Answered on 2/6/2017 by Jose from Technical Support
Displaying question 1

Do you have a question about this product? 

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat