This item is now discontinued. View the direct replacement: EW-36003-04 or browse similar items
Electrothermal Heating Mantle for 100 ml Flask, 3.31" H x 562" OD, 60 Watts, 115 V
Representative image only.
Electrothermal Item # EW-36103-02
SPECS & DESCRIPTION
Specifications & Description
  • Max Flask Size (mL)100
  • Max temperature (° F)842
  • Max temperature (° C)450
  • Power (VAC)115
  • Power (watts)60
  • DescriptionHeating Mantle for 100 ml Flask, 3.31" H x 562" OD, 60 Watts, 115 V
More About this Item
Unimantles incorporate a flexible heating element allowing for efficient heat transfer and fast boiling times. Ceramic fiber thermal insulation concentrates the heat on the glassware and keeps the polypropylene case stable and cool. Maximum temperature is 842degF (450degC). Include support rod clamp (for 1/2" rod), non-slip feet, and a 6½"-ft (2-m) two-prong line cord. All models require 115 VAC power