This item is now discontinued. Please browse similar items
Sper Scientific 840007 Analog Radiation Meter; 0.1 to 10 mR/hr, 0.1 to 100 mSv/hr
Representative image only.
Sper Scientific Mfr # 840007 – Item # EW-40451-00
Measure beta, gamma, and X-rays in mR/hr and µSv/hr.
SPECS & DESCRIPTION
Specifications & Description
  • Range0.1 to 10 mR/hr; 0.1 to 100 mSv/hr
  • DetectorGeiger-Müller (GM) tube
  • FunctionAudible alarm (20 mR/hr), low battery indication
  • BatteryOne 9 V
  • Depth (in)1
  • Width (in)3 1/4
  • Height (in)6
  • DescriptionAnalog Radiation Meter; 0.1 to 10 mR/hr, 0.1 to 100 mSv/hr
More About this Item
Choose model 40451-00 for scanning work areas, checking decontamination efforts, or monitoring areas of potential exposure. Use model 40451-10 in situations where radiation levels are higher than trace. Both models include one 9 V battery.