This item is now discontinued. Please browse similar items
Vertical float switch; polypropylene/silicone Sku4330121
Representative image only.
Item # EW-43301-21
  • Mount float switch without entering the tank
  • Insert float switch externally
  • Self-sealing grommet works with tank walls up to 5/32" (4 mm)
  • Reed switch reliability
SPECS & DESCRIPTION
Specifications & Description
  • Wetted Materialpolypropylene (float) & silicone (grommet)
  • Min Secific Gravity0.7
  • Max Process Temperature (° C)110
  • Min Process Temperature (° F)-22
  • Max Pressure (PSI)70
  • DescriptionVertical float switch; polypropylene/silicone
Key Features
  • Mount float switch without entering the tank
  • Insert float switch externally
  • Self-sealing grommet works with tank walls up to 5/32" (4 mm)
  • Reed switch reliability
More About this Item
Take the headache out of float-switch installation. This design allows you to mount the switch very easily. No need to enter the tank. Just slide the switch though the hole and tighten the nut. The compression grommet forms a seal on the inside of the tank.
Mount the horizontal switch for normally open or closed by simply rotating through 180°. The vertical and right-angled switches operate as normally open or closed by reversing the float.
Note: If your application exceeds the max amp rating, we recommend using a controller. See controllers sold separately.