This item is now discontinued. Please browse similar items
Nor-Lake Freezer, Plasma -30C 24 cu ft with Casters, 115/208-230V, 60Hz
Representative image only.
Nor-Lake Mfr # NSBF211WWW/8 – Item # EW-44173-12
Store your critical samples without worry
SPECS & DESCRIPTION
Specifications & Description
  • Freezer Capacity (cu ft)5.9
  • Freezer Capacity (Liters)167
  • Max Temperature - Freezer (° C)-10
  • Temperature ControlAnalog
  • Number of Doors1 solid
  • Number of Shelves1
  • Cabinet ConstructionWhite enamel
  • Power (VAC)115
  • Power (Hz)60
  • Power (amps)6
  • Power (watts)15
  • DescriptionFreezer, Plasma -30C 24 cu ft with Casters, 115/208-230V, 60Hz
Includes
power cord.
More About this Item
Store up to 768 300mL bags with precise temperature monitoring. Eight pull-out stainless steel drawers per door (drawers not available on undercounter model). Temperature is preset at -30-degrees Celsius and is continuously displayed on the microprocessor controller with LCD panel and message center. High and low temperature visual and audible alarms, power failure alarm, and remote alarm contacts you in case of environmental problems.
Control center with two levels of password protection helps eliminate changes in settings. Event logging with date and time stamp are stored on a non-volatile flash memory so that you can recall product temperatures in case of alarm.
Full size solid door features a key lock. Cabinet is constructed of white epoxy-coated galvanized steel and CFC free high density polyurethane foam insulation. Four casters (2 locking) allow for easy movement.