This item is now discontinued. Please browse similar items
Nor-Lake NSBR522WWG/0 Refrigerator, 52 cu ft with Casters, 2 Glass Doors, 115V/60Hz
Representative image only.
Nor-Lake Mfr # NSBR522WWG/0 – Item # EW-44173-26
Store temperature sensitive samples without worry
  • Visual and audible alarm with remote contacts
SPECS & DESCRIPTION
Specifications & Description
  • Freezer Capacity (cu ft)55.7
  • Freezer Capacity (Liters)1577
  • Min Temperature - Refrigerator (° F)39.2
  • Max Temperature - Refrigerator (° F)50
  • Min Temperature - Refrigerator (° C)4
  • Max Temperature - Refrigerator (° C)10
  • Temperature ControlMicroprocessor
  • Number of Doors2 glass
  • Number of Shelves6
  • AlarmYes
  • Cabinet ConstructionWhite enamel
  • DisplayLED
  • Power (VAC)230
  • Power (Hz)50
  • Power (amps)4.2
  • DescriptionRefrigerator, 52 cu ft with Casters, 2 Glass Doors, 115V/60Hz
Includes
cord with plug.
Key Features
  • Visual and audible alarm with remote contacts
More About this Item
Temperature is preset at 4-degrees Celsius and is continuously displayed on the microprocessor controller with LCD panel and message center. High and low temperature visual and audible alarms, power failure alarm, and remote alarm contacts you in case of environmental problems.
Control center with two levels of password protection helps eliminate changes in settings. Event logging with date and time stamp are stored on a non-volatile flash memory so that you can recall product temperatures in case of alarm. Integrated chart recorder and paper for easier temperature monitoring.
Two full size glass doors feature a key lock. Cabinet is constructed of white epoxy-coated galvanized steel and CFC free high density polyurethane foam insulation. Four casters (2 locking) allow for easy movement.