This item is now discontinued. We recommend the following as suggested replacements: EW-44261-03
Cole-Parmer Standard single-door refrigerator with hinged thermopane glass door
Representative image only.
Cole-Parmer Item # EW-44260-20
  • Keylocked power switches and doors allow storage of perishable, limited-access laboratory materials
SPECS & DESCRIPTION
Specifications & Description
  • Freezer Capacity (cu ft)24
  • Freezer Capacity (Liters)680
  • Min Temperature - Refrigerator (° F)34
  • Max Temperature - Refrigerator (° F)42
  • Min Temperature - Refrigerator (° C)1
  • Max Temperature - Refrigerator (° C)6
  • Temperature ControlMicroprocessor
  • Number of Doors1 glass
  • Number of Shelves8 Drawers
  • AlarmYes
  • Cabinet ConstructionWhite enamel
  • DisplayLCD
  • Power (VAC)230
  • Power (Hz)50
  • Power (amps)8.6
  • Power (watts)15
  • DescriptionStandard single-door refrigerator with hinged thermopane glass door
Key Features
  • Keylocked power switches and doors allow storage of perishable, limited-access laboratory materials
More About this Item
Refrigerators feature an easy-to-clean interior with epoxy-coated steel-wire shelves that resist most acids, solvents, and chemicals. Foamed-in-place insulation ensures uniform temperature control. Choose from glass- or solid-door models. Note: All doors are keylocked; models 44260-10, -20, -30, -40, and -50 feature keylocked power switches.
Compact Single-Door Refrigerator is ideal for laboratorys where space is limited. Keylocked door has spring-loaded cam-action closure—locks at 90° angle, self-closes with a gentle push. Unit features painted aluminum interior, four zinc-coated wire shelves, and six molded-in door shelves.
Standard Single-Door Refrigerators are available in hinged thermopane glass- or solid-door mo