Marvel Scientific 6CHR Chromatography Refrigerator, 6 cu ft, 115V
Representative image only.
Marvel Scientific Mfr # 6CHR – Item # EW-44765-40
Convenient, undercounter storage for all your chromatography needs
  • 1 year warranty parts & labor
SPECS & DESCRIPTION
Specifications & Description
  • Refrigerator Capacity (cu ft)5.6
  • Refrigerator Capacity (Liters)158
  • Min Temperature - Refrigerator (° F)32
  • Max Temperature - Refrigerator (° F)45
  • Min Temperature - Refrigerator (° C)0
  • Max Temperature - Refrigerator (° C)7
  • Number of Shelves2 drawers
  • Cabinet ConstructionSteel
  • DisplayLED
  • Power (VAC)115
  • Power (Hz)60
  • DescriptionChromatography Refrigerator, 6 cu ft, 115V
  • Warranty1 year parts & labor
1 Year Parts and LaborUL Listed.
Key Features
  • 1 year warranty parts & labor
More About this Item
This small, undercounter refrigerator is perfect for laboratories with limited space. Unit provides versatility to fit your demanding needs. Access ports and glass doors allow for easy instrument viewing and access. Each shelf is fully adjustable for a variety of configurations.
Constructed of white powder-coat painted steel with a dual layer of insulation—providing energy-saving efficiency. Electronic microprocessor-based thermostats are adjustable and maintain the temperature you desire. The powerful higher-rated BTU compressor provides rapid temperature recovery after warm items are placed within and after door openings.