This item is now discontinued. Please browse similar items
Marvel Scientific 8CRF General Purpose Undercounter Refrigerator/Freezer, 8 cu ft
Representative image only.
Marvel Scientific Mfr # 8CRF7100 – Item # EW-44765-50
Refrigerator/Freezers that are appropriate for noncritical temperature storage applications
  • Largest undercounter refrigerator on the market
SPECS & DESCRIPTION
Specifications & Description
  • Refrigerator Capacity (cu ft)8
  • Refrigerator Capacity (Liters)227
  • Min Temperature - Refrigerator (° F)33
  • Max Temperature - Refrigerator (° F)45
  • Min Temperature - Refrigerator (° C)1
  • Max Temperature - Refrigerator (° C)7
  • Number of Doors1 solid
  • Number of Shelves2 interior, 3 door
  • Power (VAC)115
  • Power (Hz)60
  • DescriptionGeneral Purpose Undercounter Refrigerator/Freezer, 8 cu ft
Includes
2 adjustable and removable wire shelves. Commercial power cord and plug, except for hazardous location (explosion-proof) models which require hard-wiring.
Key Features
  • Largest undercounter refrigerator on the market
More About this Item
Store heat-sensitive materials in these space-efficient refrigerators. All models have quiet, oversized refrigeration units to enable quick recovery from door openings. CFC-free refrigerant. Recessed thermostat enables flexible control. Doors are easily changed to left-side hinging.
General Purpose Models are ideal for basic storage of chemicals and reagents. These units are available in undercounter and large capacity models.