This item is now discontinued. Please browse similar items
Plastic Microscope Slides, 3" x 1", 144/pack
Representative image only.
Item # EW-48510-30
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionPlastic Microscope Slides, 3" x 1", 144/pack
More About this Item
These plastic microscope slides will not break or scratch. Made from optically clear plastic. Slides measure 3" x 1" and are 1-mm thick. Available in one gross (144 slides).
REVIEWS
Q&A
Do you have questions about this product?
Get answers from real customers and in-house experts with AnswerBox.
2 Questions | 2 Answers
Displaying questions 1-2
  • Q:
    Do the plastic slides have the same results as the glass ones? Do they come with a frosted edge?
    Asked on 1/17/2013 by Satvir from United Kingdom
    1 answer
    • STAFF EXPERT
      A:
      These slides are made of optically clear PVC which will give the same results as an optically clear glass slide. Unfortunately, and are not available with a frosted edge.
      Answered on 1/17/2013 by Jay from Customer Service
  • Q:
    can you tell me whether these slides are made of PVC? And if not what they are made of. Thanks
    Asked on 7/29/2011 by Anonymous
    1 answer
    • STAFF EXPERT
      A:
      These slides are made from PVC.
      Answered on 8/1/2011 by Jon from Technical Support
Displaying questions 1-2
Do you have a question about this product? 