Hach D33A1NN GLI D33 Dissolved Oxygen Controller, 1/4-DIN, 90 to 260 VAC
Representative image only.
Hach Mfr # D33A1NN – Item # EW-53031-00
NEMA 4X-rated, panel-mount housing
  • Built-in diagnostics continuously test analyzer and sensor operation
  • RS-232 output enables efficient data retrieval
  • 2 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Salinity CompensationManual
  • DescriptionGLI D33 Dissolved Oxygen Controller, 1/4-DIN, 90 to 260 VAC
  • Warranty2 year
Two year warrantyCanadian Standards AssociationUL Listed.
Key Features
  • Built-in diagnostics continuously test analyzer and sensor operation
  • RS-232 output enables efficient data retrieval
  • 2 year warranty
More About this Item
The D33 analyzer is ideal for monitoring dissolved oxygen concentrations in equalization basins, aeration basins, digestors, and final effluent. Menu-guided operation features up to six text lines to guide you through setup, operation, calibration, and test/maintenance functions. Display screen annunciators show relay status. Passcode capability restricts configuration and calibration access to authorized personnel only. Combine built-in diagnostics with relay customization to provide diagnostic alarm relay to alert you of sensor membrane, depleted electrolyte, and other sensor and analyzer failure conditions. Order junction box and connection cable (separately) to complete assembly.