This item is now discontinued. Please browse similar items

WTW 2V00-100 Optional Vario pH meter

Representative image only.
WTW Mfr # 2V00-100 – Item # EW-58908-00
Custom display inverts 180°
  • Innovative touch screen display
  • Built-in timer with alarm

SPECS & DESCRIPTION

Specifications & Description

  • Meter TypeBenchtop
  • Display typeLCD
  • Min pH (pH)-2
  • Max pH (pH)16
  • pH Resolution0.01
  • pH Accuracy±0.01 pH
  • Buffer RecognitionNIST and USA
  • CalibrationUp to 3 points
  • Temperature compensationAutomatic or manual
  • WaterproofYes
  • Width (cm)7.62
  • Height (cm)5.08
  • Width (in)3
  • Height (in)2
  • DescriptionOptional Vario pH meter

Key Features

  • Innovative touch screen display
  • Built-in timer with alarm

More About this Item

The VARIO combines handheld versatility with benchtop features. A simple finger tap on the display turns on the meter, setting up the initial display and inverting it if necessary. The multi-functional display can show temperature, the measured value, and any activated special functions simultaneously. The ergonomic, non-slip rubber housing makes it robust, comfortable, and easy to control all parameters with one hand. Battery-saving feature turns off the unit after 10 minutes of non-use. Measurements begin automatically once the sensor is immersed in the solution. Lock-in the measurement with the hold function. Additional GLP functions include ident-number and calibration alarm. MultiCal® allows for quick buffer recognition and calibration. The memory accommodates up to 50 time and date stamped readings. When the meter is not taking pH readings, it doubles as a precision laboratory clock or timer with alarm. What’s included: S7- and DIN-adapter; pH 4 and 7 buffers; and hard carrying case.

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat