Accumet CPXL15 XL15 Benchtop pH/mV Meter, 110/220 VAC
Representative image only.
Accumet Mfr # CPXL15 – Item # EW-59331-15
Windows® CE-driven, full-color touchscreen provides unmatched ease of use in operation and setup
  • USB, RS-232, and Ethernet/Internet connectivity — connect to printers, peripherals, LAN, or the Internet
  • Measure and display up to four inputs simultaneously without cross-channel interference
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Meter TypeBenchtop
  • Temperature compensationAutomatic or manual
  • DescriptionXL15 Benchtop pH/mV Meter, 110/220 VAC
  • Warranty1 year
1 Year WarrantyConformite EuropeenneISO9001:2000 Certified Supplier
Key Features
  • USB, RS-232, and Ethernet/Internet connectivity — connect to printers, peripherals, LAN, or the Internet
  • Measure and display up to four inputs simultaneously without cross-channel interference
  • 1 year warranty
More About this Item
The XL-series offers you the advantages and versatility of having a research-grade pH/mV meter in a easy-to-use instrument. It offers 1000 datapoint memory storage along with RS-232 communication. Password security protects your data from accidental deletion. pH mode accepts standard BNC glass electrodes as well as accuFet® electrodes directly, and allows for up to five-point pH calibration.