This item is now discontinued. View the direct replacement: EW-59340-35 or browse similar items

Accumet CPXL25 XL25 Benchtop pH/ISE/mV Meter, 110/220 VAC

Representative image only.
Accumet Mfr # CPXL25 – Item # EW-59331-25
Windows® CE-driven, full-color touchscreen provides unmatched ease of use in operation and setup
  • USB, RS-232, and Ethernet/Internet connectivity — connect to printers, peripherals, LAN, or the Internet
  • Measure and display up to four inputs simultaneously without cross-channel interference

SPECS & DESCRIPTION

Specifications & Description

  • Meter TypeBenchtop
  • Display typeLED
  • Min pH (pH)-2
  • Max pH (pH)20
  • pH Resolution0.001/0.01/0.1
  • pH Accuracy±0.1 pH
  • Buffer RecognitionDIN, NIST, USA, and custom
  • CalibrationUp to 5 points
  • Min mV (mV)-1999
  • Max mV (mV)1999
  • mV Resolution0.1 mV
  • mV Accuracy±0.2 1 digit
  • ISE Accuracy±0.17%
  • Min Temperature (° C)-10
  • Max Temperature (° C)110
  • Min Temperature (° F)14
  • Max Temperature (° F)230
  • Temperature compensationAutomatic or manual
  • InterfaceEthernet, SD card, RS-232, and USB
  • Width (cm)23.495
  • Height (cm)8.89
  • Width (in)9 1/4
  • Height (in)3 1/2
  • DescriptionXL25 Benchtop pH/ISE/mV Meter, 110/220 VAC

Includes

accuCap electrode 55500-50, ATC probe 94000-62, electrode support arm and bracket, and 110/220 VAC power supply.

Key Features

  • USB, RS-232, and Ethernet/Internet connectivity — connect to printers, peripherals, LAN, or the Internet
  • Measure and display up to four inputs simultaneously without cross-channel interference

More About this Item

The XL-series offers you the advantages and versatility of having a research-grade pH/ISE/mV meter in a easy-to-use instrument. It offers 1000 datapoint memory storage along with RS-232 communication. Password security protects your data from accidental deletion. pH mode accepts standard BNC glass electrodes as well as accuFet® electrodes directly, and allows for up to five-point pH calibration. ISE mode offers direct and indirect measurements, known and analyte, addition and subtraction methods.

ACCESSORIES

Add Optional Accessories

REVIEWS

Q&A

Do you have questions about this product?

Get answers from real customers and in-house experts with AnswerBox.

1 Question | 1 Answer
Displaying question 1
  • Q:

    I want to know what printer it match with accumet XL25
    Asked on 10/27/2011 by Anonymous

    1 answer

    • Staff Reviewer

      A:

      These meters can be used to print stored or live data to RJ-45 (Network) printers, or USB
      printers. Any printer which supports Windows CE (with PCL support) will be compatible.
      Compatible Printers we know so far (Maker, Model, Connection Type):
      HP LaserJet 1022N RJ-45 Network
      HP LaserJet 5100 RJ-45 Network
      HP Color LaserJet 2605DN RJ-45 Network
      HP LaserJet P2035N RJ-45 Network
      Konica Minolta BizHub C450 RJ-45 Network
      Dell 1700 USB
      HP 450 USB
      HP 950 USB
      HP 5740 USB
      HP DeskJet 460cb USB
      HP Deskjet 990CXI USB
      HP Laser Jet 4250/4350 USB
      HP LaserJet 1022 USB
      HP LaserJet P2035 USB
      HP OfficeJet H470 USB
      INCOMPATIBLE:
      Lexmark, Brother, Epson, Canon: Various Models, Various Connection Type
      Extech 2000i IRDA

      Answered on 10/27/2011 by Jen from Technical Support
Displaying question 1

Do you have a question about this product? 

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat