This item is now discontinued. We recommend the following as suggested replacements: EW-35423-01
Hach IQ125 Advanced ISFET pH Tester
Representative image only.
Hach Mfr # IQ125 – Item # EW-59541-10
Ion-specific field effect transistor (ISFET) uses a silicon chip pH sensor
  • Meter offers three-point calibration and auto-buffer recognition
  • Sensor stores dry and needs little maintenance
  • Fast response time
SPECS & DESCRIPTION
Specifications & Description
  • Display typeLCD
  • ConnectionsISFET or BNC
  • Min pH (pH)2
  • Max pH (pH)12
  • pH Resolution0.1
  • pH Accuracy±0.1 pH
  • CalibrationUp to 3 points
  • Min Temperature (° C)5
  • Max Temperature (° C)40
  • Min Temperature (° F)41
  • Max Temperature (° F)104
  • Temperature compensationAutomatic
  • WaterproofYes
  • Width (cm)2.8575
  • Height (cm)1.5875
  • Width (in)1
  • Height (in)0
  • DescriptionAdvanced ISFET pH Tester
Includes
calibrating buffers, case, and batteries.
Key Features
  • Meter offers three-point calibration and auto-buffer recognition
  • Sensor stores dry and needs little maintenance
  • Fast response time
More About this Item
These compact, waterproof pH meters can measure samples as small as one drop. The virtually unbreakable sensor eliminates breakage problems associated with fragile glass pH sensors.
ACCESSORIES
REVIEWS
Q&A
Do you have questions about this product?
Get answers from real customers and in-house experts with AnswerBox.
7 Questions | 7 Answers
Displaying questions 1-7
  • Q:
    Would this meter be a good choice for daily measuring of the pH level in a swimming pool?
    Asked on 5/18/2011 by Anonymous
    1 answer
    • Staff Reviewer
      A:
      Yes, as long as the sensor (of this or any other pocket pH tester) is rinsed off after testing this unit would work well to test swimming pool pH.
      Answered on 5/18/2011 by Jen from Technical Support
  • Q:
    Could this tester be used to test bar soap directly? Or should the soap be in solution?
    Asked on 2/16/2011 by Anonymous
    1 answer
    • Staff Reviewer
      A:
      This unit does not have a probe that would insert into the bar, and would not work well then to test it. If the soap was in solution this meter would work. Even ISFET probes with insertion type probes would have a hard time taking the reading of hard bar soap without any water or liquid to transfer ions through.
      Answered on 2/18/2011 by Jen from Technical Support
  • Q:
    Is this unit capable of measuring pH in very low conductivity, high viscosity corn syrup?
    Asked on 12/16/2010 by Anonymous from Decatur, IL
    1 answer
    • STAFF EXPERT
      A:
      Measuring pH is non-aqueous solutions is never easy to accomplish. This probe would not be the best for that application. Consider instead using a sleeve junction electrode such as 58820-59. There are other ways to help improve the quality of aqueous measurements including creating non-aqueous calibration buffers and changing the internal filling solution of your electrode to a non-aqueous solvent. Please contact us by phone for more details (contact information can be found on the "Contact Us" section of our website.
      Answered on 12/17/2010 by Aaron from Tech Support
  • Q:
    I am going to be canning acidified foods. Which meter do you recommend.
    Asked on 10/18/2010 by Anonymous
    1 answer
    • STAFF EXPERT
      A:
      This is a good choice for measuring the pH of foods. The stainless steel probe is easy to clean and won't leave any chemical residues that traditional pH probes do.
      Answered on 10/19/2010 by Mike from Tech Support
  • Q:
    What is the accuracy in the 1.8- 2.8 pH range.
    Is there another model better suited for low Ph applications?
    Asked on 10/12/2010 by Marty from Chicago, IL
    1 answer
    • STAFF EXPERT
      A:
      The range for this unit is listed as 2-12 pH. The accuracy within that full range is +/-0.1pH. The unit may be able to read below 2 pH, however, we do not have accuracy information outside of its listed range. To work with lower pH values, I suggest looking into our 59540-22. This is a meter and ISFET probe with a range from 0 to 14pH.
      Answered on 10/14/2010 by Josh from Technical Support
  • Q:
    Is this meter suitable for testing the pH of canning? Can it do hot ingredients?
    Asked on 6/22/2010 by holly from san martin, CA
    1 answer
    • Staff Reviewer
      A:
      This can be used to take the pH of semi-solids (cheese, jell-o, etc) and liquids. The highest temperature this unit can handle is 104F or 40C, warm but not hot.
      Answered on 6/23/2010 by Jen from Technical Support
  • Q:
    Is this meter suitable for direct soil pH testing, or does the soil have to be combined with water to form a slurry?
    Asked on 11/16/2009 by Anonymous
    1 answer
    • STAFF EXPERT
      A:
      We recommend that this pH probe be used with a slurry. Please see the following procedure:
      Using pH buffers calibrate the meter and electrode instructions.
      Loosen soil from the ground with a plastic scoop. Avoid touching soil with hands.
      Using the scoop remove any stones and crush any clump of soil to prevent damage to your electrode.
      Add equal portions of soil to distilled water in the jar. Be sure that the sample will allow sufficient immersion of your electrode. For example 50g of soil to 50 ml of water.
      Gently mix the soil slurry then let it stand for 10 minutes to allow the salts to dissolve in the soil.
      Dip electrode into the slurry and record the pH.
      Run test 3 times and average results.
      Please make sure to remove any stones or other particles in the soil that could damage the sensor (scratch it or crack it).
      For additional help, please contact our Application Specialists at the number listed on our website.
      Answered on 11/18/2009 by Jaime from Technical Support
Displaying questions 1-7
Do you have a question about this product? 