135P Food pan lid; full-size, cold/hot pans, double-sealing
Representative image only.
Mfr # 135P – Item # EW-63580-26
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionFood pan lid; full-size, cold/hot pans, double-sealing
  • Warranty1 Year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
Break resistant pans won't rust, dent, or bend. The nonstick surface makes cleaning easy; boxes are dishwasher safe. Choose from cold or hot pans. Cold pans have a temperature range from –40° to 212°F; hot boxes have a range from –40° to 375°F. Pans are color coded (clear for cold, amber for hot) for easy identification. Order lids separately below.