This item is now discontinued. We recommend the following as suggested replacements: EW-68456-25
Cole-Parmer Pressure Switch, 20 To 60 PSI, Normally Open Spst; 1/4" NPT(M) Brass Fitting
Representative image only.
Cole-Parmer Item # EW-68973-52
Low cost and short lead times make this switch perfect for OEM applications
  • Ideal for pneumatic and low pressure hydraulic applications
SPECS & DESCRIPTION
Specifications & Description
  • Min Pressure (PSI)20
  • Max Pressure (PSI)60
  • Wetted MaterialsBrass, Kapton®, nitrile
  • Process Connection1/4" NPT(M)
  • Battery Type100 VDC
  • Electrical Connectionsexposed terminals
  • Switch outputN.O.
  • OutputSPST
  • Media CompatibilityGases and liquids compatible with wetted materials
  • DescriptionPressure Switch, 20 To 60 PSI, Normally Open Spst; 1/4" NPT(M) Brass Fitting
Key Features
  • Ideal for pneumatic and low pressure hydraulic applications
More About this Item
These compact pressure switches are designed for OEM applications. Made economical with metal blade contacts instead of microswitches, these switches feature Kapton® diaphragms which maintain excellent physical properties over a wide temperature range, as well as offer superb chemical resistance with organic solvents.