Cole-Parmer Drum Pump, 35 GPM, 39", PP, ODP, 110 VAC, 60 Hz

Representative image only.
Cole-Parmer Item # EW-70613-00
System includes pump tube and motor—tube material options ensure chemical compatibility
  • Equipped with variable-speed control motors (except air and explosion-proof models) for added safety and control
  • Maintenance is reduced with no mechanical seals
  • Run dry capability

PRODUCT OPTIONS

$1,123.00USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Max Flow Rate (GPM)35
  • Max Flow Rate (LPM)132.49
  • Max Motor RPM (rpm)10000
  • Motor TypeODP
  • Power (VAC)110
  • Power (Hz)60
  • Wetted MaterialsPolypropylene, carbon, Hastelloy
  • Tube Length, Inlet (in)39
  • Outlet Connection1" hose barb
  • Max Viscosity (cP)1500
  • Max Temperature (° F)130
  • Max Temperature (° C)55
  • CE Regulatory ApprovalYes
  • UL(US) Regulatory ApprovalYes
  • Motor hp1
  • DescriptionDrum Pump, 35 GPM, 39", PP, ODP, 110 VAC, 60 Hz

Includes

electric motor with 16-ft power cord and pump tube, except models 70613-70, -75, -80, -85—they include an air motor and tube.

Key Features

  • Equipped with variable-speed control motors (except air and explosion-proof models) for added safety and control
  • Maintenance is reduced with no mechanical seals
  • Run dry capability

More About this Item

Drum and container pumps are engineered to safely transfer a variety of liquids from drums, tanks, and small storage vessels. Materials such as PP, PVDF, CPVC, and 316 SS provide excellent chemical resistance against corrosive liquids. Order optional dispensing nozzle from "Accessories" for added control.

ACCESSORIES

REVIEWS

Q&A

Do you have questions about this product?

Get answers from real customers and in-house experts with AnswerBox.

16 Questions | 16 Answers
Displaying questions 1-10Previous | Next »
  • Q:

    Wil this pump handle Hydrocloric acid
    Asked on 7/15/2016 by none from San Diego Ca

    1 answer

    • STAFF EXPERT

      A:

      No, this pump would not handle hydrocholoric acid.

      Answered on 8/1/2016 by Ethan from Technical Support
  • Q:

    Would this pump handle 90% sulphuric acid?
    Asked on 10/5/2015 by dave from Marina, Ca.

    1 answer

    • STAFF EXPERT

      A:

      The wetted materials are Polypropylene, carbon, and Hastelloy. These materials will not hold up to 90% sulfuric acid. I do not believe we a motorized drum pump that is compatible because they all contain metal wetted materials. You can determine the chemical compatibility for this chemical through our chemical compatibility chart that is linked under the technical resources tab.

      Answered on 10/5/2015 by Katie from Technical Support
  • Q:

    Will this pump work for pumping polymer from a huge tote into a 50 gallon drum on a sludge thickener?
    Asked on 5/4/2015 by TA

    1 answer

    • STAFF EXPERT

      A:

      This pump is designed for pumping from drums into production areas or smaller containers. It should work, provided you do not exceed the maximum viscosity of 1500cp, and your polymer does not have any chemical compatibility issues with the wetted parts.

      Answered on 5/4/2015 by Jay from Technical Support
  • Q:

    Can this pump transfer ATF Type F transmission fluid from a 55 gallon drum into 1 gallon cans?
    Asked on 9/15/2011 by Anonymous

    1 answer

    • STAFF EXPERT

      A:

      The specific chemical compatibility is not something we have on file, but I believe all the parts of the drum pump would be compatible with the transmission fluid, meaning it would work for your purpose.

      Answered on 9/19/2011 by Jack from Technical Support
  • Q:

    What is the maximum fluid temperature for this pump? Can it handle fluid temperatures up to 100 degrees Celsius (212 F) for short periods, say 5-10 minutes?
    Asked on 6/29/2011 by Anonymous

    1 answer

    • STAFF EXPERT

      A:

      The max temp for this pump is 130F; look at 70613-40 which has a rated temperture for continuous use at 190F (90C).

      Answered on 6/30/2011 by Jon from Technical Support
  • Q:

    Is this product approved for use with fruit juices.
    Asked on 6/11/2011 by Anonymous

    1 answer

    • STAFF EXPERT

      A:

      This product has not been FDA certified for use with food products.

      Answered on 6/14/2011 by Jack from Technical Support
  • Q:

    Is there a duty cycle available for this motor? I need to know how long I could run the pump on the highest setting.
    Asked on 3/18/2011 by Anonymous

    1 answer

    • STAFF EXPERT

      A:

      These pumps are intended for use emptying and dispensing from drums or totes and as a part of an ongoing process. Because of this intended use, they are not rated for a specific duty cycle. You could run the pump fairly continuously for an 8 hour period, slight breaks when moving from one drum to another, with no problems. After 700 hours of operation the brushes will probably need to be replaced.

      Answered on 3/22/2011 by Josh from Technical Support
  • Q:

    Which pump/mtarial would be suitable for 12% sodium hypochlorite?
    Asked on 1/12/2011 by Anonymous

    1 answer

    • STAFF EXPERT

      A:

      This pump, 70613-00 is a good choice for strong sodium hypochlorite. The materials that are resistant to sodium hypochlorite are available by using our chemical compatibility database under the technical library.

      Answered on 1/14/2011 by Jack from Technical Support
  • Q:

    Can this pump be used to pump an aqueous buffer that is used in a product that require no exposure to metal parts such as stainless steel. Also will pressure of the output from this pump will have to pass through a 10 inch filter assembly containing a 0.45 micron filter.
    thank you.
    Asked on 7/22/2010 by MJV from Los Angeles, CA

    1 answer

    • STAFF EXPERT

      A:

      The wetted parts for this pump are PP, carbon, and Hastelloy; thus it would not be suitable for an application that required no exposure to metal parts. All our motorized drum pumps have either SS or Hastelloy wetted parts.

      Answered on 7/27/2010 by David from Tech Support
  • Q:

    is this pump suitable for transferring solvent to High pressure reactor - solvent will be either Methanol, Methanolic Ammonia or Ethyl Acetate
    Asked on 6/30/2010 by Anonymous from Bangalore

    1 answer

    • STAFF EXPERT

      A:

      The wetted materials of this pump are polypropylene, ceramic, and hastelloy, which are compatible with your solvents. Note this is a drum transfer pump, it is not intended for high pressure applications. Please contact our technical application specialists for additional assistance (contact information can be found on the product web page).

      Answered on 7/6/2010 by David from Tech Support
Displaying questions 1-10Previous | Next »

Do you have a question about this product? 

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat