This item is now discontinued. Please browse similar items
Micropump L17483 Magenetically Driven Gear Pump, Mounts to NEMA (included - IEC is available), 3.17 gpm (12 l/m)
Representative image only.
Micropump Mfr # L17483 – Item # EW-73011-50
Expand fluid flow capabilities to 5.6 GPM (21,350 mL/min)
  • Withstand system pressures up to 1500 psi
  • Mounts to standard NEMA or IEC motors
  • Long life in aggressive environments
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionMagenetically Driven Gear Pump, Mounts to NEMA (included - IEC is available), 3.17 gpm (12 l/m)
Key Features
  • Withstand system pressures up to 1500 psi
  • Mounts to standard NEMA or IEC motors
  • Long life in aggressive environments
More About this Item
These cavity-style gear pumps handle your toughest fluid handling needs without taking up too much space. They always provide smooth pulseless delivery and leak-free operation with versions capable of withstanding system pressures up to 103 bar (1500 psi). The magnetic drive and a single O-ring seal keep the fluid securely inside the pump and potential contaminants out. Another design feature of the cavity-style gear pumps is the capability to handle higher inlet pressures. To meet your specific capacity or chemical resistance need, please contact an Application Specialist to size your specific pump.
Model 73011-50 has a variation that is ideal for applications like pipeline sampling due to a hybrid/abrasive-resistant option; contact an Application Specialist for details on this option.