Air-Operated Double-Diaphragm Drum Pump, PP/PTFE; 12 GPM
Representative image only.
Item # EW-74024-00
Safe and economical liquid transfer for manufacturing and water treatment environments
  • No sliding parts—pumps can run dry without damage
  • Variable flow rate and discharge pressure—one pump works for many applications
  • Portable and simple to install
PRODUCT OPTIONS
$789.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Power (Air)Yes
  • Max Air Inlet Pressure (bar)6.9
  • Max Air Inlet Pressure (psig)100
  • Wetted MaterialsPolypropylene; PTFE diaphragm, check balls and O-rings
  • Max Temperature (° C)180
  • DescriptionAir-Operated Double-Diaphragm Drum Pump, PP/PTFE; 12 GPM
Includes
90° elbow; bung adapter; polypropylene (PP) pumps include a 33"L PVC suction pipe; Kynar® (PVDF) pumps include a 33"L PVDF suction pipe.
Key Features
  • No sliding parts—pumps can run dry without damage
  • Variable flow rate and discharge pressure—one pump works for many applications
  • Portable and simple to install
More About this Item
Air-operated double-diaphragm drum pumps incorporate side liquid ports and use a 90° elbow (included) on the top of the drum. Self-priming pumps handle liquids with particulate matter at pressures up to 100 psi. Dead head operation allows discharge line to be closed with no damage or wear to the pump. Drum inlet connection is a 2" bung.
Minimal parts contact is excellent for use with shear-sensitive fluid. The PTFE diaphragms maintain their integrity when pumping highly aggressive fluids. Thermoplastic olefin (TPO) diaphragms maintain flexibility when used with acids or caustic fluids.