This item is now discontinued. Please browse similar items

Cole-Parmer PLX 23 086 10 Compact Solenoid Diaphragm Pumps, 66.7 mL/min; 115 VAC

Representative image only.
Cole-Parmer Item # EW-74402-20
  • Ideal where space is limited
  • Adjustable stroke frequency—vary flow rate via one-turn of the control knob
  • Epoxy-coated aluminum housing, stainless steel control panel; front gasket protects pump from external hazards

SPECS & DESCRIPTION

Specifications & Description

  • Max Flow Rate (mL/min)66.7
  • Max Flow Rate (Strokes/min)100
  • Flow Rate (mL/stroke)0.8
  • Max Pressure (PSI)71
  • Suction Lift, Dry (in H₂O)79.2
  • Suction Lift, Dry (mm of H₂O)2012
  • Power (VAC)115
  • Power (Hz)50/60
  • Power (amps)0.26
  • Wetted MaterialsPolypropylene, PTFE, Viton®
  • Max Temperature (° F)120
  • Max Temperature (° C)49
  • Height (in)6.5
  • Width (cm)11
  • Length (in)4.125
  • Height (cm)16.5
  • Width (in)4.3125
  • Length (cm)10.5
  • DescriptionCompact Solenoid Diaphragm Pumps, 66.7 mL/min; 115 VAC

Key Features

  • Ideal where space is limited
  • Adjustable stroke frequency—vary flow rate via one-turn of the control knob
  • Epoxy-coated aluminum housing, stainless steel control panel; front gasket protects pump from external hazards

More About this Item

All pumps include PVC inlet and discharge tubing (3/16" ID x 1/4" OD), and a 6-ft power cord with U.S. plug.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat