This item is now discontinued. Please browse similar items

LMI E701-297 Hazardous-duty solenoid diaphragm pump, 81.75 mL/min, 120 VAC

Representative image only.
LMI Mfr # W2T461887 – Item # EW-74510-00
  • Meets NEMA specifications for use in Class I Groups C and D; Class II Groups E, F, and G, and Class III Division I hazardous locations
  • Ideal for metering or chemical feed of hazardous materials or in hazardous locations
  • Adjustable stroke length and stroke frequency allow precise control of pump flow rates

SPECS & DESCRIPTION

Specifications & Description

  • Max Flow Rate (mL/min)81.75
  • Max Flow Rate (Strokes/min)100
  • Max Pressure (PSI)300
  • Suction Lift, Dry (in H₂O)60
  • Suction Lift, Dry (mm of H₂O)1524
  • Power (VAC)120
  • Power (Hz)50/60
  • Power (amps)1
  • Wetted Materials316 SS, Fluorofilm® (PTFE/PFA blend), PTFE
  • Inlet Connection1/4" NPT(M)
  • Outlet Connection1/4" NPT(M)
  • Repeatability±2%
  • Max Temperature (° F)122
  • Max Temperature (° C)50
  • Viscosity, Max (cP)220
  • Duty Cycle ContinuousYes
  • Height (in)9.0937
  • Width (cm)20.3
  • Length (in)9.875
  • Height (cm)23.1
  • Width (in)8
  • Length (cm)25.1
  • DescriptionHazardous-duty solenoid diaphragm pump, 81.75 mL/min, 120 VAC

Key Features

  • Meets NEMA specifications for use in Class I Groups C and D; Class II Groups E, F, and G, and Class III Division I hazardous locations
  • Ideal for metering or chemical feed of hazardous materials or in hazardous locations
  • Adjustable stroke length and stroke frequency allow precise control of pump flow rates
  • Remote control capabilities—pump accepts an external contact closure for flow rate
  • adjustment (closure must withstand 15 VDC at 2 mA)
  • Epoxy-coated cast iron housing protects internal components
  • Mechanically-simple solenoid pumps have one moving part—won’t stall.

More About this Item

Conduit connections are 1/2" NPT(F). Cord and plug not included. Order check valve assembly and replacement diaphragm (sold separately below right) for models 74510-00, -05, -10, and -15. For all other models, order service kits below right; kits include diaphragm, seal rings, check balls, and shaft seal.

Recommended minimum values—stroke length is adjustable from 0 to 100% on all models.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat