This item is now discontinued. We recommend the following as suggested replacements: EW-77914-20

Masterflex high-pressure pump, 115V.

Representative image only.
Masterflex Item # EW-77914-00
Pump system transfers fluids at pressures up to 125 psi (8.5 bar)
  • Pump flow rate: 0.9 to 170 mL/min; flow rate depends on drive rpm and tubing size.
  • Reversible motor - purge tubing before or after pumping; pump fluid in either direction.
  • Maintain speed setting while turning pump on/off.
  • Stackable ABS plastic housing.
  • 6-ft (1.8-m) line cord - 115 VAC: U.S. standard plug; 230 VAC: IEC 320/CEE22 socket. Please specify country to receive correct plug/cord set.

SPECS & DESCRIPTION

Specifications & Description

  • Control TypeVariable-Speed Analog
  • Pump SeriesL/S
  • Min RPM1
  • Max RPM100
  • Number of Channels1
  • Max Flow Rate Per Channel (mL/min)90
  • Min Flow Rate Per Channel (mL/min)0.9
  • Drive07520-50 (L/S® Precision Standard Variable-Speed Drive)
  • Pump Head77250-62 (L/S® High-Performance Pump Head)
  • Tubing95664-16, 8-ft (High-Pressure PharMed® BPT, L/S® 16HP)
  • Tubing sizes acceptedL/S® 14HP, 16HP, 15HP, 24HP, 15, 24, 35, 36
  • Drive IP ratingIP 23
  • Power (VAC)115
  • Power (amps)1.5
  • Power (Hz)50/60
  • Descriptionhigh-pressure pump, 115V.

Includes

L/S® High-Performance pump head 77250-62, 8-ft (2.4 m) of high-pressure PharMed® BPT L/S® 16HP pump tubing 95664-16, and an L/S® precision variable-speed drive 07528-30 (1 to 100 rpm).

Key Features

  • Pump flow rate: 0.9 to 170 mL/min; flow rate depends on drive rpm and tubing size.
  • Reversible motor - purge tubing before or after pumping; pump fluid in either direction.
  • Maintain speed setting while turning pump on/off.
  • Stackable ABS plastic housing.
  • 6-ft (1.8-m) line cord - 115 VAC: U.S. standard plug; 230 VAC: IEC 320/CEE22 socket. Please specify country to receive correct plug/cord set.

More About this Item

The Masterflex® L/S® High-Pressure pump system enables peristaltic fluid transfer using flexible tubing - at higher pressures. This innovative tubing pump system transfers fluids at pressure up to 125 psi (8.5 bar). Maintenance is quick and easy - simply change the tubing. Unlike ordinary high-pressure pumps, there are no gears, valves, seals or diaphragms to disassemble for cleaning, which saves you time, money, and downtime. This pump is ideal for chemical injection or sterile filtration because the fluid remains in the tubing at all times, with absolutely no contact with the pump's moving parts. The system uses Masterflex® High-Pressure PharMed® BPT pump tubing which is FDA compliant and complies with USP Class VI approval criteria.

ACCESSORIES

Add Optional Accessories

REVIEWS

Q&A

Do you have questions about this product?

Get answers from real customers and in-house experts with AnswerBox.

4 Questions | 4 Answers
Displaying questions 1-4
  • Q:

    Is this pump UL listed?
    Asked on 6/29/2012 by Rosene

    1 answer

    • STAFF EXPERT

      A:

      The pump drive contained in this system, item 07528-30, is UL 61010-1 compliant.

      Answered on 7/2/2012 by Jay from Customer Service
  • Q:

    Do you have any information about the smoothness of the outflow? Are there noticable pulses, or is the flow smooth?
    Asked on 10/18/2011 by Anonymous

    1 answer

    • Staff Reviewer

      A:

      Peristaltic pumps have a pulsed flow. The head on this drive has three rollers, so the pulses are pretty significant. Generally, the more rollers a head has the lower the pulse effect. Also, depending on your applicaiton we may be able to stack heads and connect the tubing with a "Y" and reduce pulsation by 96%. These high pressure heads require too much torque to run alone and use thicker tubing, they cannot be stacked. Another option would be to use a pulse dampener, if compatible and able.
      Please call so we can discuss your application and needs in further detail in order to make a more specific suggestion.

      Answered on 10/19/2011 by Jen from Technical Support
  • Q:

    Please advise if the masterflex pump 77914-00 with this code we have to order and is this pump is complete just to work. May I use the tubing 06409-16 with this pump?
    Asked on 3/15/2010 by Cecilia from Quito,Ecuador

    1 answer

    • STAFF EXPERT

      A:

      This is a complete pump system that comes with everything needed to work.
      Regular size 16 tubing like part number 06409-16 will not work in the head for this pump. Size 16HP will fit in this pump in either PharMed BPT (95664-16) or Norprene (06504-16).

      Answered on 3/17/2010 by Jen from Technical Support
  • Q:

    Is the pump explosion proof?
    What is the maximum discharge pressure?
    Asked on 10/29/2009 by Anonymous

    1 answer

    • STAFF EXPERT

      A:

      This unit is not explosion proof, the maximum discharge pressure is 100psi.

      Answered on 10/29/2009 by Jen from Technical Support
Displaying questions 1-4

Do you have a question about this product? 

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat