This item is now discontinued. View the direct replacement: EW-77921-75 or browse similar items
Masterflex L/S Standard digital pump system, 115/230 VAC
Representative image only.
Masterflex Item # EW-77921-40
SPECS & DESCRIPTION
Specifications & Description
  • Pump SeriesL/S
  • Control TypeVariable-Speed Digital
  • Min RPM10
  • Max RPM600
  • Number of Channels1
  • Max Flow Rate Per Channel (mL/min)1700
  • Min Flow Rate Per Channel (mL/min)28
  • Drive07523-60 (L/S® Brushless Digital Dispensing Drive)
  • Pump Head77200-62 (L/S® Easy-Load II Pump Head)
  • Tubing06429-24, 10-ft (Tygon® LFL tubing, L/S® 24)
  • Tubing sizes acceptedL/S® 13, 14, 16, 25, 17, 18
  • Drive IP ratingIP 33
  • Power (VAC)115 / 230
  • Power (amps)1.5A at 115v; 0.8A at 230v
  • Power (Hz)50/60
  • DescriptionL/S Standard digital pump system, 115/230 VAC
Two year warranty
More About this Item
Our L/S® economy digital drives offer wide flow ranges, powerful motors, and an easy-to-read digital display; display shows motor rpm or flow rate allowing you to monitor pump performance.
For increased accuracy these drives will store one calibration value per tubing size--even when power is turned off.
L/S economy digital drives also feature keypad lockout to prevent tampering, and tachometer feedback for ±0.25% speed control.
REVIEWS
REVIEW SNAPSHOT®
by PowerReviews
MasterflexL/S<reg> Standard digital pump system, 115/230 VAC
5.0
(based on 1 review)
Reviewed by 1 customer
Displaying review 1
Back to top
5.0
Effecient and functional
By 
from Fayette MO
About Me Lab Assistant
Verified Buyer
Pros
  • Accurate
  • Easy To Use
  • Well Calibrated
  • Works Properly
Cons
    Best Uses
    • In The Lab
    Comments about Masterflex L/S<reg> Standard digital pump system, 115/230 VAC:
    The pump holds the desired settings effectively for long periods of time.
    Displaying review 1
    Back to top
    Q&A
    Do you have questions about this product?
    Get answers from real customers and in-house experts with AnswerBox.
    4 Questions | 4 Answers
    Displaying questions 1-4
    • Q:
      I am getting Er 5 in my device? What does that mean? What I need to do?
      Asked on 9/26/2016 by MOUNIKA from NM - New Mexico
      1 answer
      • STAFF EXPERT
        A:
        Error 5 indicates an overload condition. To clear this you can do so by pressing any key. Make sure to check for proper tube loading
        and pump operation. If the issue persist the unit will need to be serviced and will need to be returned to Cole-Parmer to do so.
        Answered on 10/2/2016 by Jose from Technical Support
    • Q:
      I am looking for a PDF manual for this type of pump. We currently have one but can't find the manual for it. If you could email me one or a link that would be great. Thank you.
      Asked on 2/11/2016 by Jonathan from West Lebanon, NH
      1 answer
      • STAFF EXPERT
        A:
        This part number is for the pump system. The drive is part number 07523-60 and the manual is located on that product page. The part number for the pump head is 77200-62 and the manual is located on that product page.
        Answered on 2/11/2016 by Danielle from Technical Support
    • Q:
      I have an "Error 1" warning when attempting to use the pump. Can you provide any information on rectifying this problem?
      Asked on 11/25/2010 by TC from Manchester, UK
      1 answer
      • STAFF EXPERT
        A:
        The "Error 1" message is indicative of an internal error. The product likely needs to be repaired. Please contact our technical application specialists for additional assistance (contact information can be found in the "contact us" section on the web page).
        Answered on 11/30/2010 by Jack from Technical Support
    • Q:
      WE ARE CONSULTANTS IN FOOD AND REFRIGERATION SYSTEMS. WE ARE WORKING ON A 10000LITERS/DAY COCONUT WATER BOTTLING PLANT IN SILVASSA IN INDIA.
      WE ARE INTERESTED IN EXTRACTING 1000LITERS OF COCONUT WATER /HOUR USING MULTIPLE SS SYRINGE PUMPS. IN CASE WE HAVE 8 UNITS WE CAN EXTRACT SAY 125 LITERS/HOUR. IN EACH INJECTION WE CAN DRAW 235 ML OF COCONUT WATER FROM EACH NUT. PLEASE INFORM US SUITABILITY OF YOUR UNITS.
      Asked on 7/18/2009 by S.MANI from MUMBAI,INDIA
      1 answer
      • STAFF EXPERT
        A:
        The 77921-40 system has been discontinued, as the drive that comes with that unit has been discontinued. However, the 77921-70 system has a flowrate of 0.28-1700mL/min that should work well for your application. Please contact our technical application specialists for additional assistance (contact information can be found on the product web page).
        Answered on 7/20/2009 by Jen from Technical Support
    Displaying questions 1-4
    Do you have a question about this product? 