This item is now discontinued. View the direct replacement: EW-77924-65 or browse similar items
Masterflex Computer-Compatible Pump System with Easy-Load II pump head, 115/230 VAC
Representative image only.
Masterflex Item # EW-77924-30
SPECS & DESCRIPTION
Specifications & Description
  • Control TypeVariable-Speed Digital
  • Pump SeriesL/S
  • Min RPM10
  • Max RPM600
  • Number of Channels1
  • Max Flow Rate Per Channel (mL/min)1700
  • Min Flow Rate Per Channel (mL/min)28
  • Drive07550-30 (L/S® Computer-Compatible Digital Drive)
  • Pump Head77200-62 (L/S® Easy-Load II Pump Head)
  • Tubing06429-24, 10-ft (Tygon® LFL tubing, L/S® 24)
  • Tubing sizes acceptedL/S® 13, 14, 16, 25, 17, 18
  • Drive IP ratingIP 23
  • Power (VAC)115 / 230
  • Power (amps)1.5A at 115v; 0.8A at 230v
  • Power (Hz)50/60
  • DescriptionComputer-Compatible Pump System with Easy-Load II pump head, 115/230 VAC
Two year warranty
ACCESSORIES
Add Optional Accessories