This item is now discontinued. We recommend the following as suggested replacements: EW-77962-20
Masterflex I/P Variable Speed Modular Pump System, 20-650 RPM, 115V.
Representative image only.
Masterflex Item # EW-77962-00
SPECS & DESCRIPTION
Specifications & Description
  • Control TypeVariable-Speed Analog
  • Pump SeriesI/P
  • Min RPM20
  • Max RPM650
  • Number of Channels1
  • Max Flow Rate Per Channel (LPM)8
  • Min Flow Rate Per Channel (LPM)0.2
  • Max Flow Rate Per Channel (mL/min)8000
  • Min Flow Rate Per Channel (mL/min)20
  • Drive07591-00 (I/P® Variable-Speed Modular Drive)
  • Pump Head77601-10 (I/P® Easy-Load Pump Head)
  • Tubing06429-73, 10-ft (Tygon® LFL tubing, I/P® 73)
  • Tubing sizes acceptedI/P® 26, 73, 82
  • Drive IP ratingIP 22
  • Power (VAC)115
  • Power (amps)3.0
  • Power (Hz)50/60
  • DescriptionI/P Variable Speed Modular Pump System, 20-650 RPM, 115V.
REVIEWS
Q&A
Do you have questions about this product?
Get answers from real customers and in-house experts with AnswerBox.
8 Questions | 8 Answers
Displaying questions 1-8
  • Q:
    What is the maximum suction head, e.g. the maximum difference in elevation between the pump centerline and the end of the suction line when pumping water (I assume this is tubing-specific).
    Asked on 5/2/2013 by Jim
    1 answer
    • STAFF EXPERT
      A:
      Yes, it is tubing (and pump head) specific. This system comes with a small segment of 06440-73 tubing though. Please note as suction lift increases or feet of head/back pressure increase - the flow will decrease. So, the "maximum" is the point before the flow stops completely. Maximum suction lift is 29' (when working with water) for this tubing and head.
      Answered on 5/3/2013 by Jen from Technical Support
  • Q:
    What is the max and min size tubing this masterflex tubing can take?
    Asked on 3/4/2013 by tom
    1 answer
    • STAFF EXPERT
      A:
      This system uses the 77601-10 pump head which accept tubing sizes I/P 26, I/P 73, I/P 82.
      Answered on 3/4/2013 by Jen from Technical Support
  • Q:
    What is the margin of error or allowable error for this pump? For example, if set at 7.0 L/min should I expect 6.8-7.2L/min? 6.5-7.5L/min? or 6-8L/min?
    Asked on 12/11/2012 by Jordan from New York
    1 answer
    • STAFF EXPERT
      A:
      There are so many variable that effect flowrates that accuracy is not given in such a way.
      Back pressure, head height, suction lift, viscosity, temperature, and age, size and composition of the tubing - are all factors that effect flowrate. We can only guarantee then the speed of the drive.
      Answered on 12/13/2012 by Jen from Technical Support
  • Q:
    What is the maximum output pressure for this pump?
    Asked on 3/31/2011 by Anonymous
    1 answer
    • Staff Reviewer
      A:
      The maximum pressure of this system, as is with the Tygon tubing it comes with, is 40 psi. Please contact our technical application specialists for additional assistance (contact information can be found in the "contact us" section on the web page).
      Answered on 4/4/2011 by Jen from Technical Support
  • Q:
    Is this product Intrinsically Safe? If so, for what zone, division,ect.
    Asked on 10/19/2010 by Anonymous
    1 answer
    • Staff Reviewer
      A:
      No, the drive in this system is not rated for hazardous locations. We do offer a hazardous duty drive, please see part number 07583-50. The hazardous-duty motor is UL listed for use in Class 1, Group C and D, Division 1 locations. We also offer an air powered model 07589-30 which is ATEX Zone 2 approved (rated to EEx II 3 G c IIC T6).
      These are not complete systems, only drives.
      Please contact our technical application specialists for additional assistance (contact information can be found in the "contact us" section on the web page).
      Answered on 10/20/2010 by Jen from Technical Support
  • Q:
    What is the min and max volume per pump or pulse.
    Asked on 9/22/2010 by Anonymous
    1 answer
    • Staff Reviewer
      A:
      This system comes with the I/P size 73 tubing, the lowest drive setting is 20 rpm, and at that setting the flow rate is 0.2 LPM. 0.2/20=0.01L or 10mL per revolution.
      You could also change the pump head and tubing on this drive to accept other tubing sizes.
      Please contact our technical application specialists for additional assistance (contact information can be found in the "contact us" section on the web page).
      Answered on 9/24/2010 by Jen from Technical Support
  • Q:
    Need to purchase the brushes for the motor for this pump. Cannot find the part number for a replacement brush set.
    Asked on 4/20/2010 by Anonymous
    1 answer
    • Staff Reviewer
      A:
      Please use part number 07553-03 for the replacement cap and brush set.
      Answered on 4/21/2010 by Jen from Technical Support
  • Q:
    Hi, Is this system available for use with European voltage (220-240V) and with CE marking?
    Thanks
    Asked on 8/13/2009 by Scanners from Ireland
    1 answer
    • STAFF EXPERT
      A:
      Yes, please use part number 77962-07 for the 220V, CE marked version of this system.
      Answered on 8/14/2009 by Jen from Technical Support
Displaying questions 1-8
Do you have a question about this product? 