This item is now discontinued. We recommend the following as suggested replacements: EW-77962-30
Masterflex Variable Speed Wash-down Modular Pump System, 20-650 RPM, 115V.
Representative image only.
Masterflex Item # EW-77962-10
SPECS & DESCRIPTION
Specifications & Description
  • Control TypeVariable-Speed Analog
  • Pump SeriesI/P
  • Min RPM20
  • Max RPM650
  • Number of Channels1
  • Max Flow Rate Per Channel (LPM)8
  • Min Flow Rate Per Channel (LPM)0.2
  • Max Flow Rate Per Channel (mL/min)8000
  • Min Flow Rate Per Channel (mL/min)20
  • Drive07591-10 (I/P® Variable-Speed Wash-Down Modular Drive)
  • Pump Head77601-10 (I/P® Easy-Load Pump Head)
  • Tubing06429-73, 10-ft (Tygon® LFL tubing, I/P® 73)
  • Tubing sizes acceptedI/P® 26, 73, 82
  • Drive IP ratingIP 56
  • Power (VAC)115
  • Power (amps)3.0
  • Power (Hz)50/60
  • DescriptionVariable Speed Wash-down Modular Pump System, 20-650 RPM, 115V.